Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVW-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130925.1 | NP_001124397.1 | 2 | 2205 | ||
NM_014590.3 | NP_055405.3 | 1 | 2487 | |||
GenBank mRNA | AF072506.2 | - | 2 | 2205 | ||
AF072508.1 | - | 1 | 558 | |||
AF208161.1 | - | 1 | 2373 | |||
AF506835.1 | - | 1 | 1468 | |||
AF513360.1 | - | 1 | 1502 | |||
BC137381.1 | - | 1 | 2104 | 9021379 | ||
GQ919057.1 | - | 1 | 1444 |
Gene Symbol: ERVW-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG421046.1 | - | 1 | 14 |
Gene Symbol: ERVW-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG421049.1 | - | 1 | 14 |
Gene Symbol: ERVW-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG421056.1 | - | 1 | 14 |
Gene Symbol: ERVW-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG421066.1 | - | 1 | 14 |
Gene Symbol: ERVW-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022306.1 | - | 1 | 1871 |
Gene Symbol: ERVW-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG421044.1 | - | 1 | 15 |