Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144822.1 | NP_001138294.1 | 4 | 787 | ||
NM_001779.2 | NP_001770.1 | 4 | 787 | |||
NR_026665.1 | 4 | 787 | ||||
XM_017002869.1 | XP_016858358.1 | 4 | 787 | |||
GenBank mRNA | AK291980.1 | - | 4 | 720 | ||
BC005930.1 | - | 4 | 733 | 4285827 | ||
BT019816.1 | - | 4 | 666 | |||
BT019817.1 | - | 4 | 666 | |||
CR456939.1 | - | 4 | 666 | |||
D28586.1 | - | 2 | 318 | |||
X06296.1 | - | 4 | 678 | |||
Y00636.1 | - | 4 | 675 |