Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278785.1 | NP_001265714.1 | 2 | 273 | ||
NM_001278786.1 | NP_001265715.1 | 2 | 297 | |||
NM_014596.5 | NP_055411.1 | 2 | 282 | |||
NM_170783.3 | NP_740753.1 | 1 | 398 | |||
NR_103864.1 | 2 | 282 | ||||
GenBank mRNA | AF024616.1 | - | 1 | 367 | ||
AF024617.1 | - | 1 | 367 | |||
AF230337.1 | - | 1 | 122 | |||
AF238320.1 | - | 1 | 291 | |||
AF238321.1 | - | 2 | 243 | |||
AK225874.1 | - | 1 | 273 | |||
BC010898.1 | - | 2 | 263 | 4247252 | ||
BC050608.1 | - | 1 | 293 | 6151023 | ||
BG503963.1 | - | 2 | 275 | |||
BM798114.1 | - | 2 | 213 | |||
BQ690150.1 | - | 1 | 307 | |||
CR457109.1 | - | 1 | 113 | |||
DN996443.1 | - | 2 | 165 | |||
HY071776.1 | - | 1 | 398 |