Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278785.1 | NP_001265714.1 | 2 | 217 | ||
NM_001278786.1 | NP_001265715.1 | 2 | 241 | |||
NM_014596.5 | NP_055411.1 | 2 | 226 | |||
NM_170783.3 | NP_740753.1 | 1 | 342 | |||
NR_103864.1 | 2 | 226 | ||||
GenBank mRNA | AF024616.1 | - | 1 | 311 | ||
AF024617.1 | - | 1 | 311 | |||
AF230337.1 | - | 1 | 66 | |||
AF238320.1 | - | 1 | 235 | |||
AF238321.1 | - | 2 | 187 | |||
AK225874.1 | - | 1 | 219 | |||
BC010898.1 | - | 2 | 207 | 4247252 | ||
BC050608.1 | - | 1 | 237 | 6151023 | ||
BG503963.1 | - | 2 | 219 | |||
BM798114.1 | - | 2 | 157 | |||
BQ690150.1 | - | 1 | 251 | |||
CR457109.1 | - | 1 | 57 | |||
DN996443.1 | - | 2 | 109 | |||
HY071776.1 | - | 1 | 342 |