Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 3 | 506 | ||
NM_001144894.1 | NP_001138366.1 | 3 | 782 | |||
NM_001144895.1 | NP_001138367.1 | 5 | 638 | |||
NM_001144896.1 | NP_001138368.1 | 4 | 842 | |||
NM_001144897.1 | NP_001138369.1 | 5 | 914 | |||
NM_001144899.1 | NP_001138371.1 | 5 | 431 | |||
NM_021155.3 | NP_066978.1 | 5 | 914 | |||
NR_026692.1 | 4 | 1014 | ||||
XM_005272472.4 | XP_005272529.1 | 5 | 531 | |||
GenBank mRNA | AF290886.1 | - | 5 | 856 | ||
AK293089.1 | - | 5 | 870 | |||
AK304190.1 | - | 5 | 417 | |||
AK308595.1 | - | 2 | 457 | |||
AK313585.1 | - | 5 | 915 | |||
AY042221.1 | - | 5 | 847 | |||
AY042222.1 | - | 5 | 847 | |||
AY042223.1 | - | 5 | 571 | |||
AY042225.1 | - | 4 | 775 | |||
AY042226.1 | - | 3 | 715 | |||
AY042227.1 | - | 4 | 439 | |||
AY042229.1 | - | 4 | 947 | |||
AY042230.1 | - | 3 | 875 | |||
AY042231.1 | - | 3 | 737 | |||
AY042232.1 | - | 3 | 875 | |||
BC110615.2 | - | 3 | 882 | 40032459 | ||
M98457.1 | - | 5 | 888 |