Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014601.3 | NP_055416.2 | 2 | 623 | ||
GenBank mRNA | AF181263.1 | - | 1 | 411 | ||
AF454952.1 | - | 1 | 428 | |||
AK074701.1 | - | 2 | 545 | |||
AK296907.1 | - | 3 | 515 | |||
AK315548.1 | - | 2 | 547 | |||
BC014445.1 | - | 2 | 534 | 4908085 |