Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098402.1 | NP_001091872.1 | 3 | 1246 | ||
NM_001098403.1 | NP_001091873.1 | 3 | 1246 | |||
NM_001320729.1 | NP_001307658.1 | 2 | 1181 | |||
NM_001320731.1 | NP_001307660.1 | 4 | 1400 | |||
NM_020727.4 | NP_065778.3 | 2 | 1181 | |||
XM_005261121.3 | XP_005261178.1 | 3 | 1586 | |||
XM_011529588.2 | XP_011527890.1 | 4 | 1491 | |||
XM_011529590.2 | XP_011527892.1 | 4 | 1800 | |||
XM_017028360.1 | XP_016883849.1 | 3 | 1824 | |||
XM_017028361.1 | XP_016883850.1 | 3 | 1194 | |||
GenBank mRNA | AB033053.1 | - | 2 | 1135 | ||
AB041014.1 | - | 3 | 1216 | |||
AB041015.1 | - | 3 | 1216 | |||
AK300291.1 | - | 2 | 1142 | |||
BC063290.1 | - | 2 | 1142 | 30348821 | ||
CR933655.1 | - | 4 | 1379 |