Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257389.1 | NP_001244318.1 | 5 | 782 | ||
NM_001257390.1 | NP_001244319.1 | 5 | 525 | |||
NM_001257391.1 | NP_001244320.1 | 4 | 520 | |||
NM_001257392.1 | NP_001244321.1 | 4 | 321 | |||
NM_001257400.1 | NP_001244329.1 | 4 | 350 | |||
NM_001257401.1 | NP_001244330.1 | 4 | 490 | |||
NM_001267698.1 | NP_001254627.1 | 5 | 572 | |||
NM_001780.5 | NP_001771.1 | 5 | 576 | |||
GenBank mRNA | AF508304.1 | - | 4 | 318 | ||
AK311893.1 | - | 5 | 447 | |||
AL582859.3 | - | 4 | 395 | |||
BC002349.2 | - | 5 | 464 | 2819554 | ||
BC013017.2 | - | 5 | 420 | 3010594 | ||
BE407235.1 | - | 4 | 519 | |||
BG478842.1 | - | 5 | 526 | |||
BG766593.1 | - | 4 | 349 | |||
BQ434569.1 | - | 491 | ||||
BT007073.1 | - | 4 | 379 | |||
BT020137.1 | - | 4 | 379 | |||
BT020138.1 | - | 4 | 379 | |||
BU167765.1 | - | 5 | 583 | |||
BU190521.1 | - | 4 | 515 | |||
CD250685.1 | - | 5 | 637 | |||
CN296813.1 | - | 5 | 539 | |||
CN484590.1 | - | 4 | 390 | |||
CR542096.1 | - | 4 | 379 | |||
EB386351.1 | - | 5 | 511 | |||
KF998086.1 | - | 4 | 379 | |||
M58485.1 | - | 5 | 434 | |||
M59907.1 | - | 5 | 429 | |||
S93788.1 | - | 4 | 403 | |||
X07982.1 | - | 5 | 406 |