Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 17 | 2685 | ||
NM_014140.3 | NP_054859.2 | 17 | 2871 | |||
XM_005246631.2 | XP_005246688.1 | 17 | 2809 | |||
XM_005246632.1 | XP_005246689.1 | 17 | 2861 | |||
XM_017004228.1 | XP_016859717.1 | 17 | 2674 | |||
GenBank mRNA | AF082179.1 | - | 16 | 2641 | ||
AF432223.1 | - | 17 | 2681 | |||
AK000117.1 | - | 13 | 1541 | |||
AK023780.1 | - | 17 | 2811 | |||
AL122076.1 | - | 15 | 2105 | |||
BC016482.1 | - | 16 | 2632 | 4341673 | ||
BC029994.1 | - | 14 | 2067 | |||
BC043341.1 | - | 17 | 2658 | 6058575 |