Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 3 | 503 | ||
NM_014140.3 | NP_054859.2 | 3 | 689 | |||
XM_005246631.2 | XP_005246688.1 | 3 | 627 | |||
XM_005246632.1 | XP_005246689.1 | 3 | 679 | |||
XM_017004228.1 | XP_016859717.1 | 3 | 488 | |||
GenBank mRNA | AF432223.1 | - | 3 | 499 | ||
BC016482.1 | - | 2 | 450 | 4341673 | ||
BC043341.1 | - | 3 | 476 | 6058575 | ||
DA856812.1 | - | 3 | 503 |