Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015720.3 | NP_056535.1 | 3 | 432 | ||
GenBank mRNA | AF219137.1 | - | 3 | 391 | ||
AL524055.3 | - | 3 | 431 | |||
AY359096.1 | - | 3 | 395 | |||
BC019330.1 | - | 3 | 407 | 3502164 | ||
BC052585.1 | - | 3 | 387 | 6264322 |