Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015720.3 | NP_056535.1 | 3 | 645 | ||
GenBank mRNA | AF219137.1 | - | 3 | 604 | ||
AL524055.3 | - | 3 | 644 | |||
AY359096.1 | - | 3 | 608 | |||
BC019330.1 | - | 3 | 620 | 3502164 | ||
BC052585.1 | - | 3 | 600 | 6264322 |