Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017481.2 | NP_059509.1 | 2 | 753 | ||
XM_011520144.2 | XP_011518446.1 | 2 | 854 | |||
XM_011520145.2 | XP_011518447.1 | 2 | 823 | |||
GenBank mRNA | AF230481.1 | - | 2 | 760 | ||
BC036743.1 | - | 2 | 768 | 5167911 |