Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040059.1 | NP_001035148.1 | 5 | 881 | ||
NM_001251.2 | NP_001242.2 | 5 | 963 | |||
GenBank mRNA | AK222492.1 | - | 5 | 810 | ||
AK222514.1 | - | 5 | 809 | |||
AK290238.1 | - | 5 | 823 | |||
AK298055.1 | - | 6 | 770 | |||
AK301223.1 | - | 5 | 715 | |||
BC015557.2 | - | 5 | 835 | 3050731 | ||
BT009923.1 | - | 5 | 772 | |||
BX438711.2 | - | 5 | 720 | |||
BX647524.1 | - | 5 | 730 | |||
S57235.1 | - | 5 | 787 |