Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015721.2 | NP_056536.2 | 2 | 574 | ||
XM_005256667.4 | XP_005256724.1 | 3 | 691 | |||
XM_005256670.4 | XP_005256727.1 | 2 | 696 | |||
XM_011523910.2 | XP_011522212.1 | 3 | 952 | |||
XM_011523911.2 | XP_011522213.1 | 3 | 792 | |||
XM_011523912.2 | XP_011522214.1 | 2 | 2092 | |||
XM_011523913.2 | XP_011522215.1 | 2 | 978 | |||
XM_017024709.1 | XP_016880198.1 | 4 | 1044 | |||
GenBank mRNA | AF173856.1 | - | 2 | 478 | ||
AF177341.2 | - | 1 | 1764 | |||
AK302722.1 | - | 2 | 487 | |||
BC020062.1 | - | 2 | 504 | 4340940 | ||
BC032084.1 | - | 2 | 490 | |||
BC036024.1 | - | 2 | 478 |