Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015721.2 | NP_056536.2 | 2 | 2109 | ||
XM_005256667.4 | XP_005256724.1 | 3 | 2226 | |||
XM_005256670.4 | XP_005256727.1 | 2 | 2231 | |||
XM_011523910.2 | XP_011522212.1 | 3 | 2487 | |||
XM_011523911.2 | XP_011522213.1 | 3 | 2327 | |||
XM_011523912.2 | XP_011522214.1 | 2 | 3627 | |||
XM_011523913.2 | XP_011522215.1 | 2 | 2513 | |||
XM_017024709.1 | XP_016880198.1 | 4 | 2579 | |||
GenBank mRNA | AF173856.1 | - | 2 | 2013 | ||
AF177341.2 | - | 1 | 3299 | |||
AL080150.1 | - | 1 | 1436 | |||
BC020062.1 | - | 2 | 2039 | 4340940 | ||
BC032084.1 | - | 2 | 2025 | |||
BC036024.1 | - | 2 | 2013 |