Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015320.3 | NP_056135.2 | 8 | 1519 | ||
NM_032995.2 | NP_127462.1 | 7 | 1108 | |||
XM_005263681.1 | XP_005263738.1 | 8 | 4596 | |||
XM_005263687.3 | XP_005263744.1 | 5 | 810 | |||
XM_005263688.2 | XP_005263745.1 | 4 | 740 | |||
XM_005263689.4 | XP_005263746.1 | 5 | 655 | |||
XM_011511274.2 | XP_011509576.1 | 8 | 4596 | |||
XM_011511276.2 | XP_011509578.1 | 4 | 458 | |||
XM_011511277.1 | XP_011509579.1 | 4 | 585 | |||
GenBank mRNA | AB029035.1 | - | 7 | 1074 | ||
AB042199.1 | - | 5 | 799 | |||
AF249745.1 | - | 8 | 1520 | |||
AL137289.1 | - | 3 | 420 |