Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278632.1 | NP_001265561.1 | 3, 4 | 509 | ||
NM_001278633.1 | NP_001265562.1 | 3, 4 | 509 | |||
NM_001278634.1 | NP_001265563.1 | 3, 4 | 514 | |||
NM_001278635.1 | NP_001265564.1 | 3, 4 | 514 | |||
NM_016358.2 | NP_057442.1 | 2, 3 | 410 | |||
GenBank mRNA | AB690778.1 | - | 3, 4 | 509 | ||
AB690779.1 | - | 3, 4 | 509 | |||
AB690780.1 | - | 3, 4 | 514 | |||
AB690781.1 | - | 3, 4 | 514 | |||
AF124733.1 | - | 2, 3 | 410 | |||
BC110912.1 | - | 2, 3 | 298 | 3450161 | ||
BC136505.1 | - | 2, 3 | 398 | 9020494 | ||
BC136506.1 | - | 2, 3 | 398 | 9020495 | ||
U90306.1 | - | 1, 2 | 55 |