Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001782.2 | NP_001773.1 | 7 | 1037 | ||
XM_006716893.2 | XP_006716956.1 | 7 | 1037 | |||
XM_006716894.3 | XP_006716957.1 | 8 | 1410 | |||
XM_011518074.2 | XP_011516376.1 | 8 | 1455 | |||
XM_017015351.1 | XP_016870840.1 | 8 | 1410 | |||
GenBank mRNA | AB209587.1 | - | 5 | 2829 | ||
AF283777.2 | - | 6 | 1296 | |||
BC011734.2 | - | 7 | 948 | |||
BC030227.1 | - | 7 | 978 | 5226648 | ||
DQ174771.1 | - | 8 | 1406 | |||
M54992.1 | - | 7 | 1021 |