Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040437.2 | NP_001035527.1 | 5 | 665 | ||
NM_001040438.2 | NP_001035528.1 | 4 | 571 | |||
NM_001287482.1 | NP_001274411.1 | 5 | 676 | |||
NM_001287483.1 | NP_001274412.1 | 3 | 530 | |||
NM_001287484.1 | NP_001274413.1 | 4 | 575 | |||
NM_001287485.1 | NP_001274414.1 | 5 | 522 | |||
NM_001287486.1 | NP_001274415.1 | 5 | 519 | |||
NM_001287487.1 | NP_001274416.1 | 4 | 428 | |||
NM_001287488.1 | NP_001274417.1 | 4 | 316 | |||
GenBank mRNA | AJ249731.1 | - | 4 | 580 | ||
AJ249732.1 | - | 3 | 539 | |||
AL521625.3 | - | 5 | 651 | |||
AL556045.3 | - | 5 | 631 | |||
BC000706.1 | - | 4 | 552 | 3349773 | ||
BC017660.1 | - | 5 | 469 | 4049404 | ||
BC066330.1 | - | 4 | 570 | |||
BG109213.1 | - | 5 | 593 | |||
BG116666.1 | - | 5 | 484 | |||
BJ996767.1 | - | 3 | 530 | |||
BM690817.1 | - | 4 | 390 | |||
BQ920527.1 | - | 5 | 649 | |||
CD247791.1 | - | 5 | 538 | |||
CD249104.1 | - | 4 | 586 | |||
HY060373.1 | - | 4 | 314 |