Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037281.1 | NP_001032358.1 | 3 | 755 | ||
NM_016948.2 | NP_058644.1 | 3 | 758 | |||
XM_005255977.3 | XP_005256034.1 | 3 | 783 | |||
XM_011523095.2 | XP_011521397.1 | 3 | 780 | |||
XM_011523096.2 | XP_011521398.1 | 3 | 762 | |||
XM_017023261.1 | XP_016878750.1 | 3 | 726 | |||
GenBank mRNA | AB041642.1 | - | 3 | 720 | ||
AB043634.1 | - | 3 | 747 | |||
AF028827.1 | - | 1 | 324 | |||
AF252292.1 | - | 3 | 796 | |||
AF265565.1 | - | 3 | 729 | |||
AJ277095.1 | - | 3 | 718 | |||
BC015626.2 | - | 3 | 735 | 4844008 |