Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015987.4 | NP_057071.2 | 3 | 501 | ||
GenBank mRNA | AF117615.1 | - | 3 | 384 | ||
AF167473.1 | - | 3 | 306 | |||
AK289877.1 | - | 3 | 465 | |||
BC016277.2 | - | 3 | 336 | 3907035 | ||
BI550915.1 | - | 3 | 505 | |||
BT007294.1 | - | 3 | 306 | |||
CF126276.1 | - | 3 | 502 |