Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077196.1 | NP_001070664.1 | 4 | 381 | ||
NM_001077197.1 | NP_001070665.1 | 8 | 911 | |||
NM_001077358.1 | NP_001070826.1 | 6 | 621 | |||
NM_016953.3 | NP_058649.3 | 7 | 1880 | |||
GenBank mRNA | AB036704.1 | - | 7 | 1880 | ||
AB038041.1 | - | 8 | 911 | |||
AF281865.1 | - | 6 | 593 | |||
AJ251509.1 | - | 4 | 380 | |||
AJ278682.1 | - | 8 | 954 | |||
BC112393.1 | - | 7 | 1871 | 40068843 | ||
BC114431.1 | - | 7 | 1871 | 40068833 | ||
BX649056.1 | - | 6 | 605 | |||
HM771389.1 | - | 7 | 1562 | |||
HM771390.1 | - | 7 | 1562 | |||
HM771391.1 | - | 7 | 1562 |