Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167830.1 | NP_001161302.1 | 3 | 822 | ||
NM_016040.4 | NP_057124.3 | 3 | 822 | |||
NR_030761.1 | 4 | 893 | ||||
GenBank mRNA | AF151858.1 | - | 3 | 516 | ||
AK125882.1 | - | 4 | 593 | |||
AK293308.1 | - | 4 | 478 | |||
AK312787.1 | - | 3 | 546 | |||
AY359039.1 | - | 3 | 480 | |||
BC016365.1 | - | 3 | 540 | 4133205 | ||
BC016556.1 | - | 3 | 487 | 3048959 | ||
BC038511.1 | - | 2 | 3163 | 5262507 | ||
BC070051.1 | - | 3 | 822 | 5265257 | ||
CR936808.1 | - | 4 | 621 |