Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286643.1 | NP_001273572.1 | 9 | 1287 | ||
NM_015942.4 | NP_057026.3 | 8 | 1225 | |||
XM_006716570.3 | XP_006716633.1 | 8 | 1183 | |||
XM_011517054.2 | XP_011515356.1 | 7 | 892 | |||
GenBank mRNA | AF132946.1 | - | 7 | 947 | ||
AK001801.1 | - | 6 | 929 | |||
AK309002.1 | - | 9 | 1284 | |||
BC012995.2 | - | 8 | 1180 | 3529518 |