Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27221
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
5
650
NM_001281458.1
NP_001268387.1
6
589
NM_001281459.1
NP_001268388.1
5
650
NM_001281460.1
NP_001268389.1
6
1053
NM_015945.11
NP_057029.8
5
721
NM_173073.3
NP_775096.1
5
721
NM_173179.3
NP_775271.1
5
916
XM_011528831.1
XP_011527133.1
5
899
XM_011528832.1
XP_011527134.1
5
1193
XM_011528833.1
XP_011527135.1
5
717
XM_011528834.1
XP_011527136.1
5
721
XM_011528836.1
XP_011527138.1
5
802
XM_011528837.1
XP_011527139.1
5
636
XM_017027856.1
XP_016883345.1
5
1616
XM_017027857.1
XP_016883346.1
5
1144
XM_017027858.1
XP_016883347.1
5
1054
XM_017027859.1
XP_016883348.1
5
1421
XM_017027860.1
XP_016883349.1
6
861
XM_017027861.1
XP_016883350.1
5
883
XM_017027862.1
XP_016883351.1
5
721
GenBank mRNA
AF132949.1
-
5
658
AF455052.1
-
5
676
AK300207.1
-
5
604
AK300798.1
-
5
582
AK301022.1
-
6
531
AK315726.1
-
5
883
BC014191.2
-
5
665
4761663
BC021138.1
-
5
859
4870715
BC025277.1
-
5
917
4865888
BI831689.1
-
6
594
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00210988_m1
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline