Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198798.2 | NP_001185727.1 | 5 | 639 | ||
NM_001198799.2 | NP_001185728.1 | 6 | 684 | |||
NM_001198800.2 | NP_001185729.1 | 5 | 600 | |||
NR_045564.1 | 5 | 679 | ||||
XM_006717873.3 | XP_006717936.1 | 5 | 841 | |||
XM_011539841.2 | XP_011538143.1 | 5 | 464 | |||
XM_011539842.2 | XP_011538144.1 | 5 | 841 | |||
XM_017016291.1 | XP_016871780.1 | 5 | 464 | |||
XM_017016292.1 | XP_016871781.1 | 5 | 841 | |||
XM_017016293.1 | XP_016871782.1 | 5 | 464 | |||
XM_017016294.1 | XP_016871783.1 | 5 | 841 | |||
XM_017016295.1 | XP_016871784.1 | 5 | 464 | |||
XM_017016296.1 | XP_016871785.1 | 4 | 365 | |||
XM_017016297.1 | XP_016871786.1 | 5 | 464 | |||
XM_017016298.1 | XP_016871787.1 | 5 | 841 | |||
XM_017016299.1 | XP_016871788.1 | 5 | 464 | |||
XM_017016300.1 | XP_016871789.1 | 5 | 841 | |||
XM_017016301.1 | XP_016871790.1 | 5 | 464 | |||
XM_017016302.1 | XP_016871791.1 | 5 | 841 | |||
XM_017016303.1 | XP_016871792.1 | 4 | 365 | |||
GenBank mRNA | AF132952.1 | - | 6 | 752 | ||
AK023436.1 | - | 5 | 447 | |||
AK094170.1 | - | 6 | 518 | |||
AK096409.1 | - | 3 | 1150 | |||
AK298302.1 | - | 5 | 679 | |||
AY013290.1 | - | 6 | 752 | |||
BC012291.1 | - | 5 | 486 | 3858865 |