Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304777.1 | NP_001291706.1 | 3 | 238 | ||
NM_016041.4 | NP_057125.2 | 3 | 241 | |||
NR_130905.1 | 3 | 447 | ||||
NR_130906.1 | 3 | 241 | ||||
GenBank mRNA | AF132289.1 | - | 3 | 189 | ||
AF151859.1 | - | 3 | 178 | |||
AF208065.1 | - | 3 | 177 | |||
AF242523.1 | - | 3 | 177 | |||
BC010890.1 | - | 3 | 189 | 4280273 | ||
BG110482.1 | - | 3 | 397 | |||
CN362881.1 | - | 3 | 159 | |||
CN362882.1 | - | 3 | 159 | |||
CR457202.1 | - | 3 | 172 |