Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002269.2 | NP_001002269.1 | 1 | 115 | ||
NM_016042.3 | NP_057126.2 | 1 | 115 | |||
GenBank mRNA | AF151860.1 | - | 1 | 40 | ||
AF281132.1 | - | 1 | 91 | |||
AK289571.1 | - | 1 | 84 | |||
AK290864.1 | - | 1 | 80 | |||
BC002437.2 | - | 1 | 79 | 3346075 | ||
BC008880.2 | - | 1 | 69 | 4300826 | ||
BP237222.1 | - | 1 | 114 | |||
CF994051.1 | - | 1 | 123 | |||
DA352003.1 | - | 1 | 89 |