Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181836.5 | NP_861974.1 | 3 | 1464 | ||
GenBank mRNA | AB002450.1 | - | 1 | 251 | ||
AK027512.1 | - | 3 | 1458 | |||
AK074962.1 | - | 3 | 1284 | |||
BC019349.2 | - | 3 | 1310 | 3625342 | ||
BC107426.1 | - | 2 | 808 |