Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016065.3 | NP_057149.1 | 3 | 524 | ||
GenBank mRNA | AB049948.1 | - | 3 | 410 | ||
AF151890.1 | - | 3 | 477 | |||
AK025715.1 | - | 3 | 457 | |||
AK098291.1 | - | 3 | 470 | |||
BC021106.1 | - | 3 | 419 | 5020046 |