Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297769.1 | NP_001284698.1 | 3 | 332 | ||
NM_016067.3 | NP_057151.1 | 3 | 332 | |||
GenBank mRNA | AF151892.1 | - | 3 | 245 | ||
AK002008.1 | - | 2 | 1585 | |||
AK026620.1 | - | 3 | 299 | |||
BC005186.1 | - | 3 | 223 | 3839381 | ||
BE874586.1 | - | 3 | 174 |