Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016069.9 | NP_057153.8 | 5 | 560 | ||
GenBank mRNA | AF151894.1 | - | 5 | 467 | ||
AF349455.1 | - | 5 | 454 | |||
AK026514.1 | - | 5 | 487 | |||
BC005024.1 | - | 5 | 469 | 3955475 | ||
BG715429.1 | - | 5 | 564 |
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 31 | 3634 | ||
GenBank mRNA | AL833954.1 | - | 33 | 3788 |