Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316919.1 | NP_001303848.1 | 3 | 1419 | ||
NM_001316920.1 | NP_001303849.1 | 3 | 200 | |||
NM_001316921.1 | NP_001303850.1 | 3 | 489 | |||
NM_001316924.1 | NP_001303853.1 | 3 | 494 | |||
NM_016078.5 | NP_057162.4 | 3 | 494 | |||
GenBank mRNA | AF151906.1 | - | 3 | 494 | ||
AF223467.1 | - | 3 | 494 | |||
AK290813.1 | - | 3 | 218 | |||
BC008430.1 | - | 3 | 220 | 4292664 | ||
BX537929.1 | - | 3 | 210 | |||
BX648167.1 | - | 3 | 1419 | |||
CN390782.1 | - | 2 | 192 | |||
DA067248.1 | - | 3 | 200 | |||
DA551966.1 | - | 3 | 215 | |||
DB143840.1 | - | 3 | 462 |