Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251989.1 | NP_001238918.1 | 2, 3 | 266 | ||
NM_001251990.1 | NP_001238919.1 | 2, 3 | 190 | |||
NM_001251991.1 | NP_001238920.1 | 2, 3 | 174 | |||
NM_015895.4 | NP_056979.1 | 2, 3 | 398 | |||
XM_005249159.1 | XP_005249216.1 | 2, 3 | 437 | |||
GenBank mRNA | AF067855.1 | - | 1, 2 | 84 | ||
AK315743.1 | - | 2, 3 | 379 | |||
BC005185.1 | - | 2, 3 | 321 | 4101771 | ||
BC005389.1 | - | 2, 3 | 373 | 3996265 | ||
BF977182.1 | - | 389 | ||||
BG777134.1 | - | 2, 3 | 146 | |||
BM699599.1 | - | 2, 3 | 172 | |||
CB992796.1 | - | 2, 3 | 285 |