Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 13 | 1915 | ||
XM_011517056.2 | XP_011515358.1 | 14 | 2721 | |||
XM_011517058.2 | XP_011515360.1 | 13 | 2576 | |||
XM_011517059.1 | XP_011515361.1 | 13 | 1843 | |||
XM_011517060.2 | XP_011515362.1 | 14 | 2703 | |||
XM_011517061.2 | XP_011515363.1 | 14 | 2060 | |||
XM_011517062.1 | XP_011515364.1 | 13 | 1915 | |||
XM_011517063.2 | XP_011515365.1 | 13 | 2558 | |||
XM_011517064.2 | XP_011515366.1 | 14 | 2720 | |||
XM_011517065.1 | XP_011515367.1 | 13 | 1979 | |||
XM_011517068.2 | XP_011515370.1 | 13 | 1808 | |||
XM_011517069.2 | XP_011515371.1 | 8 | 1902 | |||
XM_011517070.2 | XP_011515372.1 | 14 | 2725 | |||
XM_011517072.2 | XP_011515374.1 | 7 | 1160 | |||
XM_011517073.2 | XP_011515375.1 | 7 | 1164 | |||
XM_011517074.1 | XP_011515376.1 | 6 | 1030 | |||
XM_017013470.1 | XP_016868959.1 | 13 | 2527 | |||
XM_017013471.1 | XP_016868960.1 | 14 | 2664 | |||
GenBank mRNA | AB196635.1 | - | 13 | 1916 | ||
AK091433.1 | - | 13 | 1915 | |||
BC037288.1 | - | 14 | 1950 | 5266568 | ||
CR936831.1 | - | 7 | 1108 |