Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 16 | 3794 | ||
XM_011517056.2 | XP_011515358.1 | 17 | 4612 | |||
XM_011517058.2 | XP_011515360.1 | 16 | 4467 | |||
XM_011517059.1 | XP_011515361.1 | 16 | 3734 | |||
XM_011517060.2 | XP_011515362.1 | 17 | 4594 | |||
XM_011517061.2 | XP_011515363.1 | 17 | 3951 | |||
XM_011517062.1 | XP_011515364.1 | 16 | 3806 | |||
XM_011517063.2 | XP_011515365.1 | 16 | 4449 | |||
XM_011517064.2 | XP_011515366.1 | 17 | 4599 | |||
XM_011517065.1 | XP_011515367.1 | 16 | 3870 | |||
XM_011517068.2 | XP_011515370.1 | 16 | 3687 | |||
XM_011517069.2 | XP_011515371.1 | 11 | 3793 | |||
XM_011517070.2 | XP_011515372.1 | 17 | 4616 | |||
XM_011517072.2 | XP_011515374.1 | 10 | 3051 | |||
XM_011517073.2 | XP_011515375.1 | 10 | 3055 | |||
XM_011517074.1 | XP_011515376.1 | 9 | 2921 | |||
XM_017013470.1 | XP_016868959.1 | 16 | 4406 | |||
XM_017013471.1 | XP_016868960.1 | 17 | 4555 | |||
GenBank mRNA | AB196635.1 | - | 16 | 3691 | ||
AF131744.1 | - | 4 | 1449 | |||
BC037288.1 | - | 17 | 3829 | 5266568 | ||
BC106912.1 | - | 4 | 1360 | |||
BC106913.1 | - | 4 | 1360 | |||
CR936831.1 | - | 10 | 2987 |