Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 7 | 774 | ||
XM_011517056.2 | XP_011515358.1 | 8 | 1580 | |||
XM_011517058.2 | XP_011515360.1 | 7 | 1435 | |||
XM_011517059.1 | XP_011515361.1 | 7 | 702 | |||
XM_011517060.2 | XP_011515362.1 | 8 | 1562 | |||
XM_011517061.2 | XP_011515363.1 | 8 | 919 | |||
XM_011517062.1 | XP_011515364.1 | 7 | 774 | |||
XM_011517063.2 | XP_011515365.1 | 7 | 1417 | |||
XM_011517064.2 | XP_011515366.1 | 8 | 1579 | |||
XM_011517065.1 | XP_011515367.1 | 7 | 838 | |||
XM_011517068.2 | XP_011515370.1 | 7 | 667 | |||
XM_011517069.2 | XP_011515371.1 | 2 | 761 | |||
XM_011517070.2 | XP_011515372.1 | 8 | 1584 | |||
XM_017013470.1 | XP_016868959.1 | 7 | 1386 | |||
XM_017013471.1 | XP_016868960.1 | 8 | 1523 | |||
GenBank mRNA | AB196635.1 | - | 7 | 775 | ||
AK091433.1 | - | 7 | 774 | |||
BC037288.1 | - | 8 | 809 | 5266568 |