Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015913.3 | NP_056997.1 | 1, 2 | 1179 | ||
NR_046405.1 | 1, 2 | 1179 | ||||
NR_046406.1 | 1, 2 | 1179 | ||||
GenBank mRNA | AF131758.1 | - | 1, 2 | 214 | ||
AF543416.1 | - | 1, 2 | 383 | |||
AK056169.1 | - | 1, 2 | 1179 | |||
AK075409.1 | - | 1, 2 | 177 | |||
AY358982.1 | - | 1, 2 | 210 | |||
BC001493.2 | - | 1, 2 | 180 | 3506109 | ||
BC008913.2 | - | 1, 2 | 209 | 3659407 | ||
BC008953.2 | - | 1, 2 | 158 | 2959502 | ||
DA277685.1 | - | 1, 2 | 227 | |||
FY213077.1 | - | 1, 2 | 116 |