Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 14 | 1825 | ||
NM_001313982.1 | NP_001300911.1 | 14 | 1813 | |||
NM_024011.3 | NP_076916.2 | 14 | 1816 | |||
NM_033529.3 | NP_277071.2 | 14 | 1786 | |||
NR_132739.1 | 14 | 1700 | ||||
NR_132740.1 | 14 | 1739 | ||||
GenBank mRNA | AF067518.1 | - | 14 | 1449 | ||
AF067519.1 | - | 14 | 1565 | |||
AF067520.1 | - | 14 | 1562 | |||
AF067521.1 | - | 14 | 1535 | |||
AF067522.1 | - | 14 | 1574 | |||
AF067523.1 | - | 14 | 1488 | |||
AK303451.1 | - | 14 | 1557 | |||
BC014464.1 | - | 6 | 547 | |||
BC094827.1 | - | 6 | 580 | |||
BC110905.1 | - | 9 | 777 | 6151813 | ||
U04819.1 | - | 7 | 889 | |||
U07704.1 | - | 14 | 1597 | |||
U07705.1 | - | 14 | 1591 |