Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137602.2 | NP_001131074.1 | 7 | 1073 | ||
NM_001301833.1 | NP_001288762.1 | 9 | 853 | |||
NM_001301852.1 | NP_001288781.1 | 7 | 632 | |||
NM_015955.3 | NP_057039.1 | 8 | 1142 | |||
NR_126032.1 | 6 | 922 | ||||
NR_126034.1 | 6 | 555 | ||||
XM_011532892.2 | XP_011531194.1 | 9 | 790 | |||
XM_011532896.2 | XP_011531198.1 | 6 | 525 | |||
XM_017004255.1 | XP_016859744.1 | 9 | 763 | |||
GenBank mRNA | AB041018.1 | - | 8 | 1142 | ||
AB041019.1 | - | 8 | 791 | |||
AF132961.1 | - | 8 | 840 | |||
AF363446.1 | - | 8 | 1059 | |||
AF529368.1 | - | 8 | 725 | |||
AK057760.1 | - | 6 | 544 | |||
AK290753.1 | - | 9 | 809 | |||
AK297128.1 | - | 7 | 784 | |||
AL161966.1 | - | 2 | 114 | |||
BC018733.1 | - | 8 | 1107 | 4851517 | ||
BC070046.1 | - | 9 | 882 | 5273070 | ||
BC094681.1 | - | 8 | 746 | 6205399 | ||
CD250945.1 | - | 6 | 619 |