Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199282.2 | NP_001186211.2 | 53 | 8193 | ||
NM_006726.4 | NP_006717.2 | 54 | 8185 | |||
XM_005263372.3 | XP_005263429.1 | 54 | 8386 | |||
XM_005263373.2 | XP_005263430.1 | 54 | 8428 | |||
XM_005263374.3 | XP_005263431.1 | 53 | 8352 | |||
XM_005263375.3 | XP_005263432.1 | 53 | 8337 | |||
XM_011532434.2 | XP_011530736.1 | 54 | 8370 | |||
XM_017008872.1 | XP_016864361.1 | 53 | 8395 | |||
XM_017008873.1 | XP_016864362.1 | 14 | 2110 | |||
XM_017008874.1 | XP_016864363.1 | 14 | 1816 | |||
GenBank mRNA | AB208838.1 | - | 29 | 3866 | ||
AF216648.2 | - | 54 | 8185 | |||
AF217149.1 | - | 7 | 777 | |||
AF467287.1 | - | 53 | 8193 | |||
AK026483.1 | - | 7 | 738 | |||
AK122920.1 | - | 17 | 2169 | |||
CR749633.1 | - | 14 | 1998 | |||
M83822.1 | - | 36 | 5621 |