Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015965.6 | NP_057049.5 | 1 | 90 | ||
GenBank mRNA | AF261134.1 | - | 1 | 521 | ||
AF286697.1 | - | 1 | 68 | |||
AK293859.1 | - | 1 | 72 | |||
BC000589.2 | - | 1 | 69 | |||
BC009189.2 | - | 1 | 77 | 3957082 | ||
BG705222.1 | - | 1 | 79 | |||
BG724220.1 | - | 1 | 565 | |||
BM839920.1 | - | 1 | 90 | |||
BP230223.1 | - | 1 | 70 |