Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253.3 | NP_001244.1 | 14 | 2305 | ||
GenBank mRNA | AB007892.2 | - | 14 | 2077 | ||
AK128737.1 | - | 14 | 2113 | |||
AK299738.1 | - | 14 | 2032 | |||
BC001568.1 | - | 14 | 2094 | 3452352 | ||
U86753.1 | - | 14 | 1995 |