Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015973.4 | NP_057057.2 | 4, 5 | 486 | ||
GenBank mRNA | BC030241.1 | - | 4, 5 | 404 | 5115887 | |
M77140.1 | - | 3, 4 | 180 |