Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT7 SIRNA Assays ›
Gene Symbol:
SEPT7
Gene Name:
septin 7
Gene Aliases:
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location:
Chr.7: 35800986 - 35915763 on Build GRCh38
UniGene ID:
Hs.191346
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2742
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011553.3
NP_001011553.2
2
361
NM_001788.5
NP_001779.3
3
364
XM_006715806.3
XP_006715869.1
3
312
XM_011515656.2
XP_011513958.1
3
312
XM_011515661.2
XP_011513963.1
3
312
XM_017012855.1
XP_016868344.1
3
296
XM_017012856.1
XP_016868345.1
3
304
XM_017012857.1
XP_016868346.1
3
301
XM_017012858.1
XP_016868347.1
3
132
XM_017012859.1
XP_016868348.1
2
129
XM_017012860.1
XP_016868349.1
3
126
XM_017012861.1
XP_016868350.1
3
123
XM_017012862.1
XP_016868351.1
2
127
XM_017012863.1
XP_016868352.1
3
670
GenBank mRNA
AB073389.1
-
3
131
AB209677.1
-
3
99
AK223225.1
-
3
131
AK223241.1
-
3
131
AK290545.1
-
2
202
AK298453.1
-
3
198
AK300601.1
-
3, 5
101
BC025987.2
-
2
128
4823810
BC067264.1
-
2
107
6503495
BC093640.1
-
3
130
7939485
BC093642.1
-
3
130
7939487
BC107718.1
-
2
309
BI669760.1
-
3
342
BP280069.1
-
3
335
BQ229116.1
-
2
292
BQ232357.1
-
3
106
BX571755.1
-
2
129
DC414394.1
-
2
364
DQ232879.1
-
2
112
KF305319.1
-
3
102
S72008.1
-
3
131
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
mitotic nuclear division
regulation of embryonic cell shape
protein heterooligomerization
cilium morphogenesis
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
structural molecule activity
protein binding
GTP binding
identical protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0