Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016034.4 | NP_057118.1 | 4 | 533 | ||
NR_051967.1 | 5 | 676 | ||||
NR_051968.1 | 5 | 731 | ||||
NR_051969.1 | 4 | 736 | ||||
NR_051970.1 | 4 | 678 | ||||
XM_006717136.3 | XP_006717199.1 | 5 | 657 | |||
GenBank mRNA | AB055388.1 | - | 4 | 457 | ||
AF151849.1 | - | 4 | 457 | |||
AL555185.3 | - | 5 | 601 | |||
BC004905.2 | - | 1 | 28 | |||
BC008017.1 | - | 4 | 469 | 3534388 | ||
BC013108.2 | - | 4 | 477 | 3924671 | ||
BQ723448.1 | - | 5 | 663 | |||
BU169388.1 | - | 4 | 594 | |||
BX362434.2 | - | 4 | 527 |