Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016037.3 | NP_057121.2 | 6, 7 | 659 | ||
XM_011541555.2 | XP_011539857.1 | 5, 6 | 593 | |||
GenBank mRNA | AF067802.1 | - | 6, 7 | 622 | ||
AF151852.1 | - | 6, 7 | 637 | |||
AK023587.1 | - | 4, 5 | 1528 | |||
AK291900.1 | - | 6, 7 | 589 | |||
AK308377.1 | - | 5, 6 | 594 | |||
CX163866.1 | - | 1, 2 | 25 |