Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016094.3 | NP_057178.2 | 4 | 366 | ||
NR_135149.1 | 4 | 446 | ||||
GenBank mRNA | AF059618.1 | - | 4 | 342 | ||
AF125096.1 | - | 4 | 352 | |||
AY542158.1 | - | 4 | 311 | |||
BC001228.1 | - | 4 | 345 | |||
BC022206.1 | - | 4 | 350 | |||
BC046131.1 | - | 4 | 328 | 4443942 | ||
BC093077.1 | - | 4 | 356 | 30372667 | ||
BI792102.1 | - | 4 | 374 | |||
BQ216242.1 | - | 1 | 109 | |||
CN483677.1 | - | 4 | 397 | |||
DA811158.1 | - | 4 | 365 |