Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 3 | 493 | ||
NM_001267708.1 | NP_001254637.1 | 4 | 537 | |||
NM_001267709.1 | NP_001254638.1 | 2 | 291 | |||
NM_016096.3 | NP_057180.1 | 2 | 378 | |||
GenBank mRNA | AF125099.1 | - | 2 | 214 | ||
AF275809.1 | - | 2 | 259 | |||
AK027802.1 | - | 2 | 1602 | |||
AK290477.1 | - | 2 | 262 | |||
BC015925.1 | - | 1 | 135 | 3909510 | ||
BC068524.1 | - | 2 | 262 | 5263977 | ||
BG704587.1 | - | 2 | 296 | |||
BI669563.1 | - | 4 | 492 | |||
BI755311.1 | - | 3 | 401 | |||
DB231109.1 | - | 2 | 291 |